Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncoa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010881.2 | NP_035011.1 | 16 - 17 | 3937 | 75 | |
GenBank mRNA | AK035922.1 | - | 1 - 2 | 261 | 75 | |
BC068177.1 | - | 13 - 14 | 3245 | 30628581 | 75 | |
BC080866.1 | - | 16 - 17 | 3700 | 30532122 | 75 | |
U56920.1 | - | 15 - 16 | 3495 | 75 | ||
U64828.1 | - | 16 - 17 | 3850 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |