Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gipr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080815.1 | NP_001074284.1 | 2 - 3 | 227 | 90 | |
GenBank mRNA | AK146552.1 | - | 2 - 3 | 228 | 90 | |
BC120671.1 | - | 1 - 2 | 131 | 40129594 | 90 | |
BC120673.1 | - | 1 - 2 | 131 | 40129596 | 90 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |