Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039106.3 | NP_001034195.2 | 2 - 3 | 1097 | 92 | |
NM_176845.5 | NP_789815.3 | 2 - 3 | 1097 | 92 | ||
GenBank mRNA | AK170271.1 | - | 3 - 4 | 811 | 92 | |
DQ272481.1 | - | 2 - 3 | 1096 | 92 | ||
DQ272482.1 | - | 2 - 3 | 1100 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |