Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdkn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028222.1 | NP_082498.1 | 2 - 3 | 194 | 88 | |
GenBank mRNA | AK010426.1 | - | 2 - 3 | 152 | 88 | |
AK033341.1 | - | 2 - 3 | 158 | 88 | ||
AK146312.1 | - | 2 - 3 | 110 | 88 | ||
BC049694.1 | - | 2 - 3 | 178 | 6705916 | 88 | |
BY084534.1 | - | 2 - 3 | 193 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Protein Phosphatase Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |