Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubxn2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145441.3 | NP_663416.1 | 1 - 2 | 176 | 103 | |
GenBank mRNA | AK031821.1 | - | 1 - 2 | 177 | 103 | |
AK041556.1 | - | 1 - 2 | 213 | 103 | ||
AK134666.1 | - | 1 - 2 | 209 | 103 | ||
AK161940.1 | - | 1 - 2 | 316 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Inventoried Probe spans exons |