Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fhit | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AA797426.1 | - | 1 - 2 | 101 | 81 | |
AF047699.1 | - | 4 - 5 | 596 | 81 | ||
AF055573.1 | - | 462 | 81 | |||
BC012662.1 | - | 2 - 3 | 321 | 4036815 | 81 | |
CJ053929.1 | - | 355 | 81 | |||
CO040033.1 | - | 4 - 5 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |