Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emcn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163522.1 | NP_001156994.1 | 10 - 11 | 914 | 77 | |
NM_016885.2 | NP_058581.2 | 9 - 10 | 875 | 77 | ||
GenBank mRNA | AB034693.1 | - | 10 - 11 | 867 | 77 | |
AF060883.1 | - | 9 - 10 | 810 | 77 | ||
AK002616.1 | - | 9 - 10 | 857 | 77 | ||
AK029348.1 | - | 881 | 77 | |||
AK164422.1 | - | 795 | 77 | |||
BC003706.1 | - | 10 - 11 | 875 | 3485788 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |