Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Odf2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162538.1 | NP_001156010.1 | 15 - 16 | 1920 | 75 | |
NM_001162539.1 | NP_001156011.1 | 13 - 14 | 1632 | 75 | ||
NM_025714.4 | NP_079990.2 | 14 - 15 | 1627 | 75 | ||
GenBank mRNA | AK014630.1 | - | 14 - 15 | 1629 | 75 | |
AK016726.1 | - | 15 - 16 | 1902 | 75 | ||
AK220159.1 | - | 4 - 5 | 3104 | 75 | ||
BC020075.1 | - | 13 - 14 | 1601 | 3979963 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |