Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300010994.mm.1 |
NM_001172055.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300010994.mm.1 |
NM_001172055.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300010994.mm.1 |
NM_027208.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300010994.mm.1 |
NM_027208.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300010994.mm.1 |
TC0300001365.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300010993.mm.1 |
NM_001172055.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300010993.mm.1 |
NM_001172055.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300010993.mm.1 |
NM_027208.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300010993.mm.1 |
NM_027208.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300010993.mm.1 |
TC0300001365.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300010995.mm.1 |
NM_001172055.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300010995.mm.1 |
NM_001172055.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300010995.mm.1 |
NM_027208.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300010995.mm.1 |
NM_027208.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300010995.mm.1 |
TC0300001365.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300010996.mm.1 |
NM_001172055.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300010996.mm.1 |
NM_001172055.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300010996.mm.1 |
NM_027208.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300010996.mm.1 |
NM_027208.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300010996.mm.1 |
TC0300001365.mm.1 |
MTA TC |