Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700011077.mm.1 |
TC0700001280.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700011077.mm.1 |
NM_001040085.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700011077.mm.1 |
NM_001040087.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700011077.mm.1 |
NM_001040088.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700011077.mm.1 |
NM_001289583.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700011077.mm.1 |
NM_001289584.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700011077.mm.1 |
NM_001289585.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700011077.mm.1 |
NM_001289586.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700011077.mm.1 |
NM_031394.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700011079.mm.1 |
TC0700001280.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700011079.mm.1 |
NM_001040085.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700011079.mm.1 |
NM_001040087.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700011079.mm.1 |
NM_001040088.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700011079.mm.1 |
NM_001289583.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700011079.mm.1 |
NM_001289584.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700011079.mm.1 |
NM_001289585.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700011079.mm.1 |
NM_001289586.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700011079.mm.1 |
NM_031394.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700011074.mm.1 |
TC0700001280.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700011074.mm.1 |
NM_001040085.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700011074.mm.1 |
NM_001040087.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700011074.mm.1 |
NM_001040088.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700011074.mm.1 |
NM_001289583.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700011074.mm.1 |
NM_001289584.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700011074.mm.1 |
NM_001289585.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700011074.mm.1 |
NM_001289586.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700011074.mm.1 |
NM_031394.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700011075.mm.1 |
TC0700001280.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700011075.mm.1 |
NM_001040085.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700011075.mm.1 |
NM_001040087.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700011075.mm.1 |
NM_001040088.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700011075.mm.1 |
NM_001289583.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700011075.mm.1 |
NM_001289584.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700011075.mm.1 |
NM_001289585.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700011075.mm.1 |
NM_001289586.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700011075.mm.1 |
NM_031394.3 |
Refseq |