Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cenpj | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014996.2 | NP_001014996.2 | 12 - 13 | 3552 | 76 | |
GenBank mRNA | AK077045.1 | - | 12 - 13 | 3535 | 76 | |
AK136260.1 | - | 2 - 3 | 139 | 76 | ||
BC092399.1 | - | 12 - 13 | 3538 | 30549055 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |