Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145514.5 | NP_663489.4 | 9 - 10 | 1904 | 78 | |
GenBank mRNA | AK075743.1 | - | 9 - 10 | 1296 | 78 | |
AK082326.1 | - | 10 - 11 | 1199 | 78 | ||
AK194190.1 | - | 2 - 3 | 237 | 78 | ||
AK201026.1 | - | 2 - 3 | 237 | 78 | ||
BC020044.1 | - | 5 - 6 | 649 | 78 | ||
BC058601.1 | - | 7 - 8 | 900 | 78 | ||
BC094233.1 | - | 6 - 7 | 788 | 78 | ||
BC139336.1 | - | 9 - 10 | 1066 | 8862358 | 78 | |
BC139337.1 | - | 9 - 10 | 1066 | 8862359 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |