Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198653.2 | NP_941055.1 | 15 - 16 | 2061 | 88 | |
GenBank mRNA | AK036182.1 | - | 15 - 16 | 2029 | 88 | |
AK043590.1 | - | 15 - 16 | 2063 | 88 | ||
AK048653.1 | - | 15 - 16 | 2062 | 88 | ||
AK050358.1 | - | 15 - 16 | 2044 | 88 | ||
AK051778.1 | - | 15 - 16 | 2022 | 88 | ||
AK078183.1 | - | 15 - 16 | 2063 | 88 | ||
AK085714.1 | - | 15 - 16 | 2059 | 88 | ||
AK088965.1 | - | 15 - 16 | 2022 | 88 | ||
AK166832.1 | - | 15 - 16 | 2044 | 88 | ||
BC027432.1 | - | 5 - 6 | 519 | 88 | ||
BC037123.1 | - | 10 - 11 | 1121 | 88 | ||
BC052403.1 | - | 15 - 16 | 2038 | 5715723 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |