Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291222.1 | NP_001278151.1 | 9 - 10 | 2238 | 87 | |
NM_010626.3 | NP_034756.2 | 9 - 10 | 2238 | 87 | ||
GenBank mRNA | BC141045.1 | - | 9 - 10 | 2239 | 9055657 | 87 |
BC145222.1 | - | 9 - 10 | 2239 | 9053773 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |