Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ticrr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029835.1 | NP_084111.1 | 10 - 11 | 2389 | 83 | |
GenBank mRNA | AK041726.1 | - | 10 - 11 | 2339 | 83 | |
AK051639.1 | - | 10 - 11 | 2352 | 83 | ||
AK054492.1 | - | 10 - 11 | 2339 | 83 | ||
AK137597.1 | - | 10 - 11 | 2394 | 83 | ||
AK157802.1 | - | 9 - 10 | 3946 | 83 | ||
BC158103.1 | - | 10 - 11 | 2353 | 9088180 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |