Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700029250.mm.1 |
NM_146191.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029250.mm.1 |
NM_146191.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029250.mm.1 |
TC0700003434.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700029246.mm.1 |
NM_146191.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029246.mm.1 |
NM_146191.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029246.mm.1 |
TC0700003434.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700029267.mm.1 |
NM_146191.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029267.mm.1 |
NM_146191.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029267.mm.1 |
TC0700003434.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700029247.mm.1 |
NM_146191.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029247.mm.1 |
NM_146191.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029247.mm.1 |
TC0700003434.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700029251.mm.1 |
NM_146191.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029251.mm.1 |
NM_146191.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029251.mm.1 |
TC0700003434.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700029252.mm.1 |
NM_146191.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029252.mm.1 |
NM_146191.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029252.mm.1 |
TC0700003434.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700029248.mm.1 |
NM_146191.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029248.mm.1 |
NM_146191.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029248.mm.1 |
TC0700003434.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700029269.mm.1 |
NM_146191.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029269.mm.1 |
NM_146191.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029269.mm.1 |
TC0700003434.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700029245.mm.1 |
NM_146191.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029245.mm.1 |
NM_146191.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029245.mm.1 |
TC0700003434.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700029270.mm.1 |
NM_146191.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029270.mm.1 |
NM_146191.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029270.mm.1 |
TC0700003434.mm.1 |
MTA TC |