Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700029283.mm.1 |
NM_146191.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029283.mm.1 |
NM_146191.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029283.mm.1 |
TC0700003434.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700029288.mm.1 |
NM_146191.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029288.mm.1 |
NM_146191.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029288.mm.1 |
TC0700003434.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700029289.mm.1 |
NM_146191.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029289.mm.1 |
NM_146191.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029289.mm.1 |
TC0700003434.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700029298.mm.1 |
NM_146191.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029298.mm.1 |
NM_146191.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029298.mm.1 |
TC0700003434.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700029279.mm.1 |
NM_146191.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029279.mm.1 |
NM_146191.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029279.mm.1 |
TC0700003434.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700029282.mm.1 |
NM_146191.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029282.mm.1 |
NM_146191.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029282.mm.1 |
TC0700003434.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700029281.mm.1 |
NM_146191.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029281.mm.1 |
NM_146191.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029281.mm.1 |
TC0700003434.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700029285.mm.1 |
NM_146191.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029285.mm.1 |
NM_146191.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029285.mm.1 |
TC0700003434.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700029296.mm.1 |
NM_146191.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029296.mm.1 |
NM_146191.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029296.mm.1 |
TC0700003434.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700029278.mm.1 |
NM_146191.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029278.mm.1 |
NM_146191.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029278.mm.1 |
TC0700003434.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700029280.mm.1 |
NM_146191.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029280.mm.1 |
NM_146191.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029280.mm.1 |
TC0700003434.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700029286.mm.1 |
NM_146191.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029286.mm.1 |
NM_146191.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029286.mm.1 |
TC0700003434.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700029299.mm.1 |
NM_146191.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029299.mm.1 |
NM_146191.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029299.mm.1 |
TC0700003434.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700029287.mm.1 |
NM_146191.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029287.mm.1 |
NM_146191.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029287.mm.1 |
TC0700003434.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700029301.mm.1 |
NM_146191.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029301.mm.1 |
NM_146191.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029301.mm.1 |
TC0700003434.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700029284.mm.1 |
NM_146191.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029284.mm.1 |
NM_146191.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029284.mm.1 |
TC0700003434.mm.1 |
MTA TC |