Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gbp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153564.2 | NP_705792.2 | 8 - 9 | 1427 | 79 | |
GenBank mRNA | AF422243.1 | - | 8 - 9 | 1409 | 79 | |
AF487898.1 | - | 6 - 7 | 1083 | 79 | ||
AK030414.1 | - | 8 - 9 | 1427 | 79 | ||
AK087083.1 | - | 1515 | 79 | |||
BC058555.1 | - | 8 - 9 | 1461 | 5293575 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |