Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ak5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081277.1 | NP_001074746.1 | 10 - 11 | 1427 | 79 | |
GenBank mRNA | AK053807.1 | - | 10 - 11 | 1427 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |