Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smyd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027188.3 | NP_081464.1 | 3 - 4 | 385 | 69 | |
GenBank mRNA | AK010447.1 | - | 3 - 4 | 419 | 69 | |
AK043988.1 | - | 3 - 4 | 395 | 69 | ||
AK087728.1 | - | 3 - 4 | 397 | 69 | ||
AK163057.1 | - | 3 - 4 | 402 | 69 | ||
AK167875.1 | - | 3 - 4 | 400 | 69 | ||
BC019663.1 | - | 3 - 4 | 387 | 69 | ||
BC038482.1 | - | 3 - 4 | 266 | 69 | ||
BC052431.1 | - | 3 - 4 | 385 | 6837012 | 69 | |
BC061485.1 | - | 3 - 4 | 368 | 5370924 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |