Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ceacam20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027839.2 | NP_082115.2 | 9 - 10 | 1607 | 145 | |
GenBank mRNA | AK018613.1 | - | 9 - 10 | 1604 | 145 | |
AK033581.2 | - | 9 - 10 | 1608 | 145 | ||
BC049361.1 | - | 9 - 10 | 1588 | 6476373 | 145 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |