Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Traf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011633.2 | NP_035763.2 | 967 | 65 | ||
GenBank mRNA | AK141097.1 | - | 6 - 7 | 928 | 65 | |
AK144694.1 | - | 918 | 65 | |||
BC012702.1 | - | 921 | 4218625 | 65 | ||
D78141.1 | - | 944 | 65 | |||
D83528.1 | - | 809 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |