Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clca1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017474.2 | NP_059502.1 | 4 - 5 | 588 | 158 | |
GenBank mRNA | AB017156.1 | - | 4 - 5 | 571 | 158 | |
AK007466.1 | - | 4 - 5 | 585 | 158 | ||
AK008659.2 | - | 4 - 5 | 585 | 158 | ||
AK033560.1 | - | 589 | 158 | |||
AK136476.1 | - | 4 - 5 | 585 | 158 | ||
BC028343.1 | - | 4 - 5 | 578 | 4217548 | 158 | |
BC116318.2 | - | 4 - 5 | 571 | 40095991 | 158 | |
BC116319.2 | - | 4 - 5 | 571 | 40095992 | 158 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons |