Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hs2st1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011828.3 | NP_035958.3 | 2 - 3 | 720 | 118 | |
GenBank mRNA | AF060178.1 | - | 2 - 3 | 398 | 118 | |
AF169243.1 | - | 2 - 3 | 387 | 118 | ||
AK088698.1 | - | 2 - 3 | 696 | 118 | ||
AK165112.1 | - | 2 - 3 | 686 | 118 | ||
BC025443.1 | - | 2 - 3 | 648 | 5068428 | 118 | |
BC059008.1 | - | 2 - 3 | 477 | 6849136 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |