Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irf9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159417.1 | NP_001152889.1 | 6 - 7 | 909 | 69 | |
NM_001159418.1 | NP_001152890.1 | 5 - 6 | 707 | 69 | ||
NM_008394.3 | NP_032420.1 | 5 - 6 | 823 | 69 | ||
GenBank mRNA | AK145911.1 | - | 4 - 5 | 575 | 69 | |
AK149742.1 | - | 4 - 5 | 574 | 69 | ||
BC005435.1 | - | 6 - 7 | 684 | 3601255 | 69 | |
BC012968.1 | - | 5 - 6 | 718 | 3257714 | 69 | |
CK788642.1 | - | 5 - 6 | 650 | 69 | ||
U51992.1 | - | 5 - 6 | 627 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |