Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polr3d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164082.1 | NP_001157554.1 | 1 - 2 | 274 | 100 | |
NM_025945.3 | NP_080221.3 | 2 - 3 | 306 | 100 | ||
GenBank mRNA | AK013165.1 | - | 1 - 2 | 191 | 100 | |
AK156587.1 | - | 2 - 3 | 258 | 100 | ||
BC016102.1 | - | 2 - 3 | 282 | 4911475 | 100 | |
CA548436.1 | - | 1 - 2 | 223 | 100 | ||
CD553533.1 | - | 2 - 3 | 288 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |