Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164220.1 | NP_001157692.1 | 1 - 2 | 122 | 68 | |
GenBank mRNA | AF220129.1 | - | 1 - 2 | 137 | 68 | |
AK162129.1 | - | 1 - 2 | 110 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |