Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rwdd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025637.3 | NP_079913.2 | 2 - 3 | 250 | 60 | |
GenBank mRNA | AK011007.1 | - | 2 - 3 | 250 | 60 | |
AK011008.1 | - | 2 - 3 | 250 | 60 | ||
AK050254.1 | - | 1 - 2 | 117 | 60 | ||
BC128288.1 | - | 2 - 3 | 249 | 60 | ||
BC150838.1 | - | 2 - 3 | 250 | 9087749 | 60 | |
BY710389.1 | - | 2 - 3 | 250 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |