Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hs6st2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077202.2 | NP_001070670.1 | 3 - 4 | 986 | 92 | |
NM_001290468.1 | NP_001277397.1 | 2 - 3 | 627 | 92 | ||
GenBank mRNA | AB024565.1 | - | 2 - 3 | 549 | 92 | |
AK043774.1 | - | 2 - 3 | 110 | 92 | ||
AK052348.1 | - | 3 - 4 | 766 | 92 | ||
AK083281.1 | - | 2 - 3 | 614 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |