Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc42bpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033285.1 | NP_001028457.1 | 33 - 34 | 4691 | 117 | |
GenBank mRNA | AK129148.1 | - | 4 - 5 | 506 | 117 | |
BC158017.1 | - | 33 - 34 | 4691 | 9007323 | 117 | |
BC158095.1 | - | 32 - 33 | 4652 | 9007382 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |