Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlgap5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144553.2 | NP_653136.2 | 2 - 3 | 530 | 108 | |
GenBank mRNA | AB076696.1 | - | 2 - 3 | 530 | 108 | |
AK036384.1 | - | 2 - 3 | 332 | 108 | ||
AK048470.1 | - | 2 - 3 | 300 | 108 | ||
AK085882.1 | - | 2 - 3 | 311 | 108 | ||
AK129032.1 | - | 2 - 3 | 276 | 108 | ||
AK135820.1 | - | 2 - 3 | 337 | 108 | ||
AK147955.1 | - | 2 - 3 | 300 | 108 | ||
AK164575.1 | - | 2 - 3 | 325 | 108 | ||
BC027273.1 | - | 2 - 3 | 270 | 108 | ||
BC058087.1 | - | 2 - 3 | 264 | 6816660 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |