Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlgap5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144553.2 | NP_653136.2 | 4 - 5 | 795 | 94 | |
GenBank mRNA | AB076696.1 | - | 4 - 5 | 795 | 94 | |
AK036384.1 | - | 4 - 5 | 597 | 94 | ||
AK048470.1 | - | 4 - 5 | 565 | 94 | ||
AK085882.1 | - | 4 - 5 | 576 | 94 | ||
AK135820.1 | - | 4 - 5 | 602 | 94 | ||
AK147955.1 | - | 4 - 5 | 565 | 94 | ||
BC058087.1 | - | 4 - 5 | 529 | 6816660 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |