Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172598.3 | NP_766186.2 | 2 - 3 | 450 | 135 | |
GenBank mRNA | AK042443.1 | - | 1 - 2 | 2063 | 135 | |
AK052690.1 | - | 2 - 3 | 426 | 135 | ||
BC063740.1 | - | 2 - 3 | 383 | 6328626 | 135 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |