Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145546.1 | NP_663521.1 | 3 - 4 | 283 | 111 | |
GenBank mRNA | AK131856.1 | - | 3 - 4 | 305 | 111 | |
AK133097.1 | - | 3 - 4 | 305 | 111 | ||
AK143935.1 | - | 3 - 4 | 305 | 111 | ||
AK145403.1 | - | 3 - 4 | 305 | 111 | ||
AK146223.1 | - | 3 - 4 | 306 | 111 | ||
AK146666.1 | - | 3 - 4 | 305 | 111 | ||
AK146910.1 | - | 3 - 4 | 305 | 111 | ||
AK151577.1 | - | 3 - 4 | 305 | 111 | ||
BC016637.1 | - | 3 - 4 | 283 | 2650779 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |