Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031388.2 | NP_113565.2 | 2 - 3 | 269 | 69 | |
GenBank mRNA | AK082829.1 | - | 2 - 3 | 196 | 69 | |
AK133417.1 | - | 2 - 3 | 273 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Detect Genomic DNA Mus musculus Probe spans exons |