Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23467997 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23467992 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23467995 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23467996 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23467993 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P314907 |
AJ003132.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P314907 |
AJ003132.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
748932 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
626319 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
146497 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
930220 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
521744 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
512788 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
539198 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
169215 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
632871 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
178492 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
611168 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
770969 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
773023 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
13500 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
441325 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
527986 |
AK016292.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
527986 |
AK017084.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
527986 |
BC125317.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
527986 |
NM_025763.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
153808 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
177780 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
142491 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P120020 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P124642 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P314907 |
AJ003132.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P314907 |
AJ003132.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P404570 |
BC050827.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P314907 |
AJ003132.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P314907 |
AJ003132.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P397418 |
AK014480.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P397418 |
AK083650.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23467975 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23467997 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23467963 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23467990 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23467977 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23467973 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23467991 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23467994 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23467964 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23467980 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23467986 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23467960 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23467970 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23467968 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23467984 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23467983 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23467974 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23467993 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23467971 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23467979 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23467988 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P124642 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P01035443 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P120020 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P05475750 |
- |
- |
Whole Mouse Genome G4122A |
A_52_P397418 |
AK014480.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P397418 |
AK083650.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P404570 |
BC050827.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P314907 |
AJ003132.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P314907 |
AJ003132.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P121567 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P139014 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P121436 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P124642 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P120020 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P04273656 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P04273654 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23467995 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23467993 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23467996 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23467997 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23467992 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467959 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467800 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467845 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467908 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467940 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467938 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467794 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467865 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467838 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467789 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467877 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467941 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467948 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467957 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467842 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467967 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467804 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467851 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467870 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467830 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467915 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467924 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467946 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467868 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467819 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467826 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467928 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467933 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467806 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467875 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467927 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467977 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467867 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467879 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467991 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467876 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467920 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467795 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467811 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467808 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467799 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467810 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467797 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467964 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467953 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467981 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467994 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467881 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467814 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467848 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467988 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467822 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467858 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467979 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467942 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467945 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467834 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467813 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467986 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467802 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467960 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467935 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467997 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467861 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467824 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467860 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467913 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467934 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467816 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467983 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467910 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467968 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467956 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467888 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467973 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467856 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467923 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467840 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467884 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467931 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467970 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467863 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467975 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467909 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467873 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467882 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467818 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467836 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467897 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467852 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467916 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467893 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467925 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467895 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467901 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467850 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467971 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467952 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467828 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467944 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467914 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467891 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467886 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467855 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467899 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23467872 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23467970 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23467918 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23467967 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23467836 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23467913 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23467981 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23467910 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23467828 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04338930 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23467956 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23467975 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23467973 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23467908 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04749443 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23467953 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04741662 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23467991 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04273659 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23467834 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23467905 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23467955 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23467959 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23467988 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23467916 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23467983 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23467994 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23467968 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04913494 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23467827 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04361379 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23467824 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04427943 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23467915 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23467964 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23467979 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23467822 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23467960 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23467917 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23467961 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23467914 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23467952 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23467977 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23467829 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04235527 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04719233 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23467830 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23467986 |
- |
- |