Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200028124.mm.1 |
NM_001164358.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200028124.mm.1 |
NM_001164358.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200028124.mm.1 |
TC0200003304.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200028125.mm.1 |
NM_001164358.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200028125.mm.1 |
NM_001164358.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200028125.mm.1 |
TC0200003304.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200028118.mm.1 |
NM_001164358.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200028118.mm.1 |
NM_001164358.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200028118.mm.1 |
NM_146118.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200028118.mm.1 |
NM_146118.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200028118.mm.1 |
TC0200003304.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200028126.mm.1 |
NM_001164358.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200028126.mm.1 |
NM_001164358.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200028126.mm.1 |
TC0200003304.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200028129.mm.1 |
NM_001164358.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200028129.mm.1 |
NM_001164358.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200028129.mm.1 |
TC0200003304.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200028119.mm.1 |
NM_001164357.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200028119.mm.1 |
NM_001164357.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200028119.mm.1 |
NM_001164358.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200028119.mm.1 |
NM_001164358.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200028119.mm.1 |
NM_146118.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200028119.mm.1 |
NM_146118.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200028119.mm.1 |
TC0200003304.mm.1 |
MTA TC |