Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030112.2 | NP_084388.2 | 6 - 7 | 907 | 82 | |
GenBank mRNA | BC117848.1 | - | 6 - 7 | 759 | 40093717 | 82 |
BC117849.1 | - | 6 - 7 | 759 | 40093724 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |