Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030559.3 | NP_085036.3 | 15 - 16 | 1555 | 79 | |
GenBank mRNA | AB056721.1 | - | 15 - 16 | 1513 | 79 | |
AK032558.1 | - | 6 - 7 | 516 | 79 | ||
AK049872.1 | - | 15 - 16 | 1512 | 79 | ||
AK162984.1 | - | 12 - 13 | 1878 | 79 | ||
AK169694.1 | - | 12 - 13 | 2094 | 79 | ||
BC003944.1 | - | 3 - 4 | 427 | 3487257 | 79 | |
BC025626.1 | - | 15 - 16 | 1521 | 5320572 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |