Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il13ra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK089687.2 | - | 10 - 11 | 1677 | 123 | |
BC003723.1 | - | 7 - 8 | 853 | 3486223 | 123 | |
EF219410.1 | - | 5 - 6 | 692 | 123 | ||
U65747.1 | - | 7 - 8 | 941 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |