Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aloxe3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011786.2 | NP_035916.2 | 1 - 2 | 167 | 80 | |
GenBank mRNA | AK141003.1 | - | 1 - 2 | 330 | 80 | |
BC116258.1 | - | 1 - 2 | 166 | 40093733 | 80 | |
BC116259.1 | - | 1 - 2 | 166 | 40093735 | 80 | |
Y14695.1 | - | 1 - 2 | 167 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |