Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polr3f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029763.3 | NP_084039.2 | 4 - 5 | 434 | 100 | |
GenBank mRNA | AK013897.1 | - | 4 - 5 | 421 | 100 | |
AK029286.1 | - | 4 - 5 | 435 | 100 | ||
AK036166.1 | - | 4 - 5 | 431 | 100 | ||
AK076538.1 | - | 4 - 5 | 461 | 100 | ||
AK144679.1 | - | 4 - 5 | 432 | 100 | ||
BC009159.1 | - | 4 - 5 | 423 | 3602180 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |