Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Specc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029936.3 | NP_001025107.1 | 11 - 12 | 2963 | 66 | |
NM_001281818.1 | NP_001268747.1 | 13 - 14 | 3180 | 66 | ||
GenBank mRNA | AK220465.1 | - | 11 - 12 | 2890 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |