Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Skap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018773.2 | NP_061243.1 | 5 - 6 | 528 | 68 | |
GenBank mRNA | AB014485.1 | - | 5 - 6 | 445 | 68 | |
AF051324.1 | - | 5 - 6 | 443 | 68 | ||
AK076000.1 | - | 5 - 6 | 473 | 68 | ||
AK144289.1 | - | 5 - 6 | 465 | 68 | ||
BC003711.1 | - | 5 - 6 | 528 | 3599914 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |