Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_180678.3 | NP_851009.2 | 8 - 9 | 1045 | 62 | |
GenBank mRNA | AK012758.2 | - | 8 - 9 | 1040 | 62 | |
AK146238.1 | - | 8 - 9 | 1022 | 62 | ||
AK165857.1 | - | 8 - 9 | 993 | 62 | ||
AK190416.1 | - | 1 - 2 | 109 | 62 | ||
AK190448.1 | - | 2 - 3 | 211 | 62 | ||
AK194283.1 | - | 2 - 3 | 187 | 62 | ||
AK196051.1 | - | 2 - 3 | 211 | 62 | ||
BC021747.1 | - | 8 - 9 | 1026 | 4192628 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |