Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamtsl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029981.1 | NP_084257.1 | 12 - 13 | 1960 | 85 | |
GenBank mRNA | AB096928.1 | - | 12 - 13 | 1960 | 85 | |
AK156991.1 | - | 12 - 13 | 1963 | 85 | ||
BC128568.1 | - | 12 - 13 | 1912 | 40097415 | 85 | |
BC129984.1 | - | 12 - 13 | 1912 | 40097418 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons |