Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc34a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011392.2 | NP_035522.2 | 10 - 11 | 1275 | 63 | |
GenBank mRNA | AB163921.1 | - | 1280 | 63 | ||
AB163922.1 | - | 10 - 11 | 1219 | 63 | ||
AK018740.2 | - | 10 - 11 | 1230 | 63 | ||
AK032618.1 | - | 3 - 4 | 366 | 63 | ||
BC020529.1 | - | 10 - 11 | 1218 | 4223484 | 63 | |
L33878.1 | - | 9 - 10 | 1202 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |