Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uimc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001293660.1 | NP_001280589.1 | 3 - 4 | 344 | 89 | |
NM_011307.2 | NP_035437.1 | 3 - 4 | 339 | 89 | ||
NR_121592.1 | 3 - 4 | 344 | 89 | |||
GenBank mRNA | AK052704.1 | - | 3 - 4 | 340 | 89 | |
AK078206.1 | - | 3 - 4 | 343 | 89 | ||
AK083342.1 | - | 3 - 4 | 280 | 89 | ||
AK160926.1 | - | 3 - 4 | 345 | 89 | ||
AK161177.1 | - | 3 - 4 | 343 | 89 | ||
BC037092.1 | - | 3 - 4 | 300 | 89 | ||
BC040808.1 | - | 3 - 4 | 295 | 4038778 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |