Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gle1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028923.3 | NP_083199.1 | 4 - 5 | 682 | 91 | |
GenBank mRNA | AK016671.1 | - | 4 - 5 | 664 | 91 | |
AK028183.1 | - | 4 - 5 | 666 | 91 | ||
AK133888.1 | - | 4 - 5 | 634 | 91 | ||
AK145748.1 | - | 4 - 5 | 658 | 91 | ||
AK150230.1 | - | 4 - 5 | 657 | 91 | ||
AK161659.1 | - | 4 - 5 | 665 | 91 | ||
BC026797.1 | - | 4 - 5 | 662 | 3990668 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |