Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btbd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145900.1 | NP_001139372.1 | 3 - 4 | 429 | 108 | |
NM_201646.2 | NP_964008.2 | 2 - 3 | 511 | 108 | ||
GenBank mRNA | AK146991.1 | - | 2 - 3 | 343 | 108 | |
BC031195.1 | - | 2 - 3 | 511 | 5059510 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |